Assay Details
| Gene Symbol: LOC4340564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788359.2 | XP_015643845.1 | 3 - 4 | 1928 | 77 | |
| GenBank mRNA | AK070606.1 | - | 3 - 4 | 1725 | 77 | |
More Information
Set Membership: |
|

LOC4340564
Genomic Map