Assay Details
| Gene Symbol: LOC4340743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788424.2 | XP_015643910.1 | 1 - 2 | 163 | ||
| GenBank mRNA | AK066417.1 | - | 1 - 2 | 267 | 163 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788424.2 transcripts.
Set Membership: |
|

LOC4340743
Genomic Map