Assay Details
| Gene Symbol: LOC4340743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788424.2 | XP_015643910.1 | 6 - 7 | 1109 | 75 | |
| GenBank mRNA | AK066417.1 | - | 6 - 7 | 743 | 75 | |
More Information
Set Membership: |
|

LOC4340743
Genomic Map