Assay Details
| Gene Symbol: LOC4340743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788424.2 | XP_015643910.1 | 8 - 9 | 1709 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4340743| Gene Symbol: LOC4340743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788424.2 | XP_015643910.1 | 8 - 9 | 1709 | 133 | |
Set Membership: |
|