Assay Details
| Gene Symbol: LOC4341460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787688.2 | XP_015643174.1 | 2 - 3 | 1007 | 109 | |
| XM_015787689.2 | XP_015643175.1 | 2 - 3 | 1002 | 109 | ||
| XM_015787690.2 | XP_015643176.1 | 1 - 2 | 628 | 109 | ||
More Information
Set Membership: |
|

LOC4341460
Genomic Map