Assay Details
| Gene Symbol: LOC4341460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787688.2 | XP_015643174.1 | 3 - 4 | 1167 | 80 | |
| XM_015787689.2 | XP_015643175.1 | 3 - 4 | 1162 | 80 | ||
| XM_015787690.2 | XP_015643176.1 | 2 - 3 | 788 | 80 | ||
More Information
Set Membership: |
|

LOC4341460
Genomic Map