Assay Details
| Gene Symbol: LOC4341520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787703.2 | XP_015643189.1 | 1 - 2 | 131 | 84 | |
| GenBank mRNA | AK069720.1 | - | 1 - 2 | 74 | 84 | |
| AK104175.1 | - | 1 - 2 | 77 | 84 | ||
| AK104562.1 | - | 1 - 2 | 76 | 84 | ||
| AK104605.1 | - | 1 - 2 | 76 | 84 | ||
| BT130373.1 | - | 1 - 2 | 75 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015787703.2 transcripts.
Set Membership: |
|

LOC4341520
Genomic Map