Assay Details
| Gene Symbol: LOC9267494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785690.2 | XP_015641176.1 | 2 - 3 | 345 | 99 | |
| XM_015785692.2 | XP_015641178.1 | 2 - 3 | 371 | 99 | ||
| GenBank mRNA | AK071675.1 | - | 2 - 3 | 338 | 99 | |
More Information
Set Membership: |
|

LOC9267494
Genomic Map