Assay Details
| Gene Symbol: LOC4341683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785793.2 | XP_015641279.1 | 1 - 2 | 594 | 87 | |
| XM_015785794.2 | XP_015641280.1 | 2 - 3 | 524 | 87 | ||
| XM_015785795.2 | XP_015641281.1 | 1 - 2 | 594 | 87 | ||
| GenBank mRNA | AK122015.1 | - | 1 - 2 | 262 | 87 | |
More Information
Set Membership: |
|

LOC4341683
Genomic Map