Assay Details
| Gene Symbol: LOC4341956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788078.2 | XP_015643564.1 | 1 - 2 | 378 | 116 | |
| GenBank mRNA | AK071376.1 | - | 1 - 2 | 183 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788078.2 transcripts.
Set Membership: |
|

LOC4341956
Genomic Map