Assay Details
| Gene Symbol: LOC4341956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788078.2 | XP_015643564.1 | 2 - 3 | 810 | 81 | |
| XM_015788079.2 | XP_015643565.1 | 2 - 3 | 622 | 81 | ||
| XM_015788080.2 | XP_015643566.1 | 2 - 3 | 584 | 81 | ||
| GenBank mRNA | AJ512642.1 | - | 2 - 3 | 445 | 81 | |
| AK071376.1 | - | 2 - 3 | 615 | 81 | ||
More Information
Set Membership: |
|

LOC4341956
Genomic Map