Assay Details
| Gene Symbol: LOC9271614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789032.2 | XP_015644518.1 | 2 - 3 | 3125 | 96 | |
| XM_015789033.2 | XP_015644519.1 | 2 - 3 | 3088 | 96 | ||
| GenBank mRNA | AK073053.1 | - | 1 - 2 | 2662 | 96 | |
More Information
Set Membership: |
|

LOC9271614
Genomic Map