Assay Details
| Gene Symbol: LOC4342047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786175.2 | XP_015641661.1 | 1 - 2 | 396 | 66 | |
| GenBank mRNA | AB003323.1 | - | 1 - 2 | 288 | 66 | |
| AK069317.1 | - | 1 - 2 | 402 | 66 | ||
More Information
Set Membership: |
|

LOC4342047
Genomic Map