Assay Details
| Gene Symbol: LOC4342047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786175.2 | XP_015641661.1 | 5 - 6 | 670 | 72 | |
| GenBank mRNA | AB003323.1 | - | 5 - 6 | 562 | 72 | |
| AK069317.1 | - | 5 - 6 | 676 | 72 | ||
More Information
Set Membership: |
|

LOC4342047
Genomic Map