Assay Details
| Gene Symbol: LOC4342450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791893.2 | XP_015647379.1 | 1 - 2 | 868 | 106 | |
| XM_015791895.2 | XP_015647381.1 | 1 - 2 | 868 | 106 | ||
| XM_015791896.2 | XP_015647382.1 | 1 - 2 | 203 | 106 | ||
| GenBank mRNA | AK065685.1 | - | 1 - 2 | 797 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791896.2 transcripts.
Set Membership: |
|

LOC4342450
Genomic Map