Assay Details
Gene Symbol: LOC4342680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791038.1 | XP_015646524.1 | 11 - 12 | 1525 | 93 | |
GenBank mRNA | AF054629.2 | - | 7 - 8 | 876 | 93 | |
AK062079.1 | - | 5 - 6 | 658 | 93 | ||
AK065213.1 | - | 11 - 12 | 1494 | 93 | ||
AK066223.1 | - | 8 - 9 | 913 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
