Assay Details
Gene Symbol: LOC4342680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791038.1 | XP_015646524.1 | 13 - 14 | 1760 | 65 | |
GenBank mRNA | AF054629.2 | - | 9 - 10 | 1111 | 65 | |
AK062079.1 | - | 7 - 8 | 893 | 65 | ||
AK065213.1 | - | 13 - 14 | 1729 | 65 | ||
AK066223.1 | - | 10 - 11 | 1148 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
