Assay Details
Gene Symbol: LOC4342680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791038.1 | XP_015646524.1 | 2 - 3 | 472 | 53 | |
GenBank mRNA | AK065213.1 | - | 2 - 3 | 441 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
