Assay Details
Gene Symbol: LOC4342680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791038.1 | XP_015646524.1 | 7 - 8 | 1106 | 150 | |
GenBank mRNA | AF054629.2 | - | 3 - 4 | 457 | 150 | |
AK062079.1 | - | 1 - 2 | 239 | 150 | ||
AK065213.1 | - | 7 - 8 | 1075 | 150 | ||
AK066223.1 | - | 4 - 5 | 494 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
