Assay Details
Gene Symbol: LOC4342680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791038.1 | XP_015646524.1 | 8 - 9 | 1193 | 114 | |
GenBank mRNA | AF054629.2 | - | 4 - 5 | 544 | 114 | |
AK062079.1 | - | 2 - 3 | 326 | 114 | ||
AK065213.1 | - | 8 - 9 | 1162 | 114 | ||
AK066223.1 | - | 5 - 6 | 581 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
