Assay Details
| Gene Symbol: LOC4342824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789234.2 | XP_015644720.1 | 3 - 4 | 362 | 93 | |
| GenBank mRNA | AK100710.1 | - | 2 - 3 | 1636 | 93 | |
| BT131175.1 | - | 1 - 2 | 77 | 93 | ||
| BT132417.1 | - | 3 - 4 | 355 | 93 | ||
More Information
Set Membership: |
|

LOC4342824
Genomic Map