Assay Details
| Gene Symbol: LOC9268104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791405.2 | XP_015646891.1 | 5 - 6 | 818 | 99 | |
| GenBank mRNA | AK108133.1 | - | 5 - 6 | 658 | 99 | |
More Information
Set Membership: |
|

LOC9268104
Genomic Map