Assay Details
Gene Symbol: LOC4343412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791089.2 | XP_015646575.1 | 17 - 18 | 3785 | 91 | |
GenBank mRNA | AK105311.1 | - | 5 - 6 | 796 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
