Assay Details
| Gene Symbol: LOC4343626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789216.2 | XP_015644702.1 | 2 - 3 | 547 | 74 | |
| GenBank mRNA | AK071983.1 | - | 2 - 3 | 412 | 74 | |
| AK107481.1 | - | 1 - 2 | 622 | 74 | ||
More Information
Set Membership: |
|

LOC4343626
Genomic Map