Assay Details
| Gene Symbol: LOC4343626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789216.2 | XP_015644702.1 | 4 - 5 | 744 | 107 | |
| GenBank mRNA | AK071983.1 | - | 4 - 5 | 609 | 107 | |
| AK107481.1 | - | 3 - 4 | 819 | 107 | ||
More Information
Set Membership: |
|

LOC4343626
Genomic Map