Assay Details
| Gene Symbol: LOC4343626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789216.2 | XP_015644702.1 | 8 - 9 | 1052 | 109 | |
| GenBank mRNA | AK071983.1 | - | 8 - 9 | 917 | 109 | |
| AK107481.1 | - | 7 - 8 | 1127 | 109 | ||
More Information
Set Membership: |
|

LOC4343626
Genomic Map