Assay Details
| Gene Symbol: LOC4343626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789216.2 | XP_015644702.1 | 9 - 10 | 1210 | 77 | |
| GenBank mRNA | AK071983.1 | - | 9 - 10 | 1075 | 77 | |
| AK107481.1 | - | 8 - 9 | 1285 | 77 | ||
More Information
Set Membership: |
|

LOC4343626
Genomic Map