Assay Details
| Gene Symbol: LOC4343803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791573.2 | XP_015647059.1 | 10 - 11 | 1771 | 78 | |
| XM_015791575.2 | XP_015647061.1 | 10 - 11 | 1771 | 78 | ||
| XM_015791576.2 | XP_015647062.1 | 9 - 10 | 1711 | 78 | ||
| GenBank mRNA | AK073674.1 | - | 10 - 11 | 1625 | 78 | |
| AK122151.1 | - | 10 - 11 | 1674 | 78 | ||
More Information
Set Membership: |
|

LOC4343803
Genomic Map