Assay Details
| Gene Symbol: LOC4343803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791573.2 | XP_015647059.1 | 12 - 13 | 1986 | 66 | |
| XM_015791575.2 | XP_015647061.1 | 12 - 13 | 1986 | 66 | ||
| XM_015791576.2 | XP_015647062.1 | 11 - 12 | 1926 | 66 | ||
| GenBank mRNA | AK122151.1 | - | 12 - 13 | 1889 | 66 | |
More Information
Set Membership: |
|

LOC4343803
Genomic Map