Assay Details
| Gene Symbol: LOC4343803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791575.2 | XP_015647061.1 | 16 - 17 | 2369 | 115 | |
| GenBank mRNA | AK122151.1 | - | 16 - 17 | 2272 | 115 | |
More Information
Set Membership: |
|

LOC4343803
Genomic Map