Assay Details
| Gene Symbol: LOC4343803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791573.2 | XP_015647059.1 | 18 - 19 | 2615 | 84 | |
| XM_015791575.2 | XP_015647061.1 | 18 - 19 | 2585 | 84 | ||
| XM_015791576.2 | XP_015647062.1 | 17 - 18 | 2555 | 84 | ||
| GenBank mRNA | AK073674.1 | - | 18 - 19 | 2439 | 84 | |
| AK122151.1 | - | 18 - 19 | 2488 | 84 | ||
More Information
Set Membership: |
|

LOC4343803
Genomic Map