Assay Details
| Gene Symbol: LOC4343803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791573.2 | XP_015647059.1 | 22 - 23 | 3365 | 141 | |
| XM_015791575.2 | XP_015647061.1 | 22 - 23 | 3335 | 141 | ||
| XM_015791576.2 | XP_015647062.1 | 21 - 22 | 3305 | 141 | ||
| GenBank mRNA | AK073674.1 | - | 22 - 23 | 3189 | 141 | |
| AK122151.1 | - | 22 - 23 | 3238 | 141 | ||
More Information
Set Membership: |
|

LOC4343803
Genomic Map