Assay Details
| Gene Symbol: LOC4343803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791573.2 | XP_015647059.1 | 23 - 24 | 3445 | 80 | |
| XM_015791575.2 | XP_015647061.1 | 23 - 24 | 3415 | 80 | ||
| XM_015791576.2 | XP_015647062.1 | 22 - 23 | 3385 | 80 | ||
| GenBank mRNA | AK073674.1 | - | 23 - 24 | 3269 | 80 | |
| AK122151.1 | - | 23 - 24 | 3318 | 80 | ||
More Information
Set Membership: |
|

LOC4343803
Genomic Map