Assay Details
| Gene Symbol: LOC4343803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791573.2 | XP_015647059.1 | 24 - 25 | 3588 | 126 | |
| XM_015791575.2 | XP_015647061.1 | 24 - 25 | 3558 | 126 | ||
| XM_015791576.2 | XP_015647062.1 | 23 - 24 | 3528 | 126 | ||
| GenBank mRNA | AK073674.1 | - | 24 - 25 | 3412 | 126 | |
More Information
Set Membership: |
|

LOC4343803
Genomic Map