Assay Details
| Gene Symbol: LOC4344106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792562.2 | XP_015648048.1 | 10 - 11 | 1238 | 93 | |
| GenBank mRNA | AK065269.1 | - | 10 - 11 | 1204 | 93 | |
More Information
Set Membership: |
|

LOC4344106
Genomic Map