Assay Details
| Gene Symbol: LOC4344154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791180.1 | XP_015646666.1 | 11 - 12 | 1404 | 74 | |
| GenBank mRNA | AK072927.1 | - | 11 - 12 | 1328 | 74 | |
| AK100911.1 | - | 11 - 12 | 1268 | 74 | ||
More Information
Set Membership: |
|

LOC4344154
Genomic Map