Assay Details
| Gene Symbol: LOC4344183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791315.2 | XP_015646801.1 | 5 - 6 | 4289 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4344183| Gene Symbol: LOC4344183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791315.2 | XP_015646801.1 | 5 - 6 | 4289 | 96 | |
Set Membership: |
|