Assay Details
| Gene Symbol: LOC4344327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791083.2 | XP_015646569.1 | 1 - 2 | 311 | 69 | |
| XM_015791084.2 | XP_015646570.1 | 1 - 2 | 303 | 69 | ||
| XM_015791085.2 | XP_015646571.1 | 1 - 2 | 333 | 69 | ||
| XM_015791086.2 | XP_015646572.1 | 1 - 2 | 317 | 69 | ||
| GenBank mRNA | AK067645.1 | - | 1 - 2 | 223 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791083.2, XM_015791084.2, XM_015791085.2, XM_015791086.2 transcripts.
Set Membership: |
|

LOC4344327
Genomic Map