Assay Details
| Gene Symbol: LOC4344327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791083.2 | XP_015646569.1 | 3 - 4 | 596 | 107 | |
| XM_015791084.2 | XP_015646570.1 | 3 - 4 | 588 | 107 | ||
| XM_015791085.2 | XP_015646571.1 | 3 - 4 | 618 | 107 | ||
| XM_015791086.2 | XP_015646572.1 | 3 - 4 | 602 | 107 | ||
| XM_015791087.2 | XP_015646573.1 | 3 - 4 | 621 | 107 | ||
| GenBank mRNA | AK067645.1 | - | 3 - 4 | 508 | 107 | |
More Information
Set Membership: |
|

LOC4344327
Genomic Map