Assay Details
| Gene Symbol: LOC4344327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791083.2 | XP_015646569.1 | 4 - 5 | 657 | 100 | |
| XM_015791084.2 | XP_015646570.1 | 4 - 5 | 649 | 100 | ||
| XM_015791087.2 | XP_015646573.1 | 4 - 5 | 682 | 100 | ||
| GenBank mRNA | AK067645.1 | - | 4 - 5 | 569 | 100 | |
More Information
Set Membership: |
|

LOC4344327
Genomic Map