Assay Details
| Gene Symbol: LOC4344327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791084.2 | XP_015646570.1 | 5 - 6 | 739 | 71 | |
| XM_015791086.2 | XP_015646572.1 | 5 - 6 | 750 | 71 | ||
| GenBank mRNA | AK067645.1 | - | 5 - 6 | 659 | 71 | |
More Information
Set Membership: |
|

LOC4344327
Genomic Map