Assay Details
| Gene Symbol: LOC4344347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789563.2 | XP_015645049.1 | 7 - 8 | 930 | 67 | |
| GenBank mRNA | AK073511.1 | - | 7 - 8 | 928 | 67 | |
| AK103992.1 | - | 3 - 4 | 268 | 67 | ||
More Information
Set Membership: |
|

LOC4344347
Genomic Map