Assay Details
| Gene Symbol: LOC4344467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795585.2 | XP_015651071.1 | 8 - 9 | 960 | 109 | |
| GenBank mRNA | AK107316.1 | - | 8 - 9 | 961 | 109 | |
More Information
Set Membership: |
|

LOC4344467
Genomic Map