Assay Details
| Gene Symbol: LOC4344938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794915.1 | XP_015650401.1 | 1 - 2 | 578 | 90 | |
| GenBank mRNA | AK073315.1 | - | 1 - 2 | 584 | 90 | |
| AK108310.1 | - | 1 - 2 | 606 | 90 | ||
| AK120546.1 | - | 1 - 2 | 584 | 90 | ||
More Information
Set Membership: |
|

LOC4344938
Genomic Map