Assay Details
| Gene Symbol: LOC4344938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794915.1 | XP_015650401.1 | 5 - 6 | 1037 | 90 | |
| GenBank mRNA | AK120546.1 | - | 5 - 6 | 1043 | 90 | |
More Information
Set Membership: |
|

LOC4344938
Genomic Map