Assay Details
| Gene Symbol: LOC4345725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794444.2 | XP_015649930.1 | 7 - 8 | 3696 | 80 | |
| GenBank mRNA | AK102479.1 | - | 2 - 3 | 236 | 80 | |
More Information
Set Membership: |
|

LOC4345725
Genomic Map