Assay Details
| Gene Symbol: LOC4345911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795481.2 | XP_015650967.1 | 8 - 9 | 3586 | 75 | |
| XM_015795482.2 | XP_015650968.1 | 8 - 9 | 2867 | 75 | ||
| GenBank mRNA | AK241274.1 | - | 8 - 9 | 2866 | 75 | |
More Information
Set Membership: |
|

LOC4345911
Genomic Map