Assay Details
| Gene Symbol: LOC4346340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792768.2 | XP_015648254.1 | 3 - 4 | 350 | 124 | |
| GenBank mRNA | AK062315.1 | - | 3 - 4 | 318 | 124 | |
More Information
Set Membership: |
|

LOC4346340
Genomic Map