Assay Details
| Gene Symbol: LOC4346355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794480.2 | XP_015649966.1 | 4 - 5 | 1289 | 143 | |
| GenBank mRNA | AK067576.1 | - | 4 - 5 | 1296 | 143 | |
More Information
Set Membership: |
|

LOC4346355
Genomic Map