Assay Details
| Gene Symbol: LOC4346586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795895.2 | XP_015651381.1 | 1 | 881 | 114 | |
| GenBank mRNA | AK240903.1 | - | 1 | 667 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795895.2 transcripts.
Set Membership: |
|

LOC4346586
Genomic Map